Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LPPR4 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524895

Catalog No. PIPA524895


Add to cart

Description

Description

This antibody is predicted to react with mouse based on sequence homology.

LPPR4 belongs to the lipid phosphate phosphatase (LPP) family. LPPs catalyze the dephosphorylation of a number of bioactive lipid mediators that regulate a variety of cell functions. This protein is specifically expressed in neurons. It is located in the membranes of outgrowing axons and has been shown to be important for axonal outgrowth during development and regenerative sprouting.
Specifications

Specifications

LPPR4
Polyclonal
PBS with 0.09% sodium azide
Q7Z2D5
LPR4, PHP1, PRG-1, PRG1, RP4-788L13.1, brain-specific phosphatidic acid phosphatase-like protein 1, plasticity related gene 1, plasticity-related gene 1 protein
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
9890
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
PLPPR4
Liquid
PLPPR4
KLH conjugated synthetic peptide between 666-695 amino acids from the C-terminal region of human LPPR4
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit