Learn More
LPPR3 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA112727
Catalog No. 01-674-854
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
A suggested positive control for this product is human brain lysate.
Phospholipid-mediated signalling on neurons provokes diverse responses such as neurogenesis, pattern formation and neurite remodelling. A recently uncovered novel set of molecules in the mammalian brain, named plasticity-related genes (PRGs) mediate lipid phosphate phosphatase activity and provide evidence for their involvement in mechanisms of neuronal plasticity.
Specifications
Specifications
LPPR3
Polyclonal
PBS with 0.08% sodium azide
Q6T4P5
LPR3, PRG-2, PRG2
Rabbit
IgG
10 Miniblots
-80° C, Avoid Freeze/Thaw Cycles
Primary
79948
ELISA, Western Blot
Unconjugated
PLPPR3
Liquid
PLPPR3
Synthetic peptide corresponding to LPPRP3.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human