Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRFN5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA520700
Description
A suggested positive control is EL4 cell lysate. PA5-20700 can be used with blocking peptide PEP-0815.
LRFN5 Polyclonal antibody specifically detects LRFN5 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blot.Specifications
LRFN5 | |
Polyclonal | |
Unconjugated | |
LRFN5 | |
AI427653; AI604817; C130061B21; C14orf146; fibronectin type III, immunoglobulin and leucine rich repeat domains 8; FIGLER8; Kiaa4208; leucine rich repeat and fibronectin type III domain containing 5; leucine-rich repeat and fibronectin type-III domain-containing protein 5; LRFN5; mKIAA4208; SALM5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
145581, 238205, 314164 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
D4A1J9, Q8BXA0, Q96NI6 | |
LRFN5 | |
A 16 amino acid peptide near the carboxy terminus of human LRFN5. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction