Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma anti-LRP-5/6 Clone: 1A12,
Mouse Monoclonal Antibody
Manufacturer: MilliporeSigma 48947550UG
Description
Specifically detects LRP-5/6 Clone: 1A12 in Human, Mouse samples, and it is validated for Immuno Blotting
LRP-5 and LRP-6 are members of the LDL receptor family involved in the Wnt signaling pathway. The peptide sequence corresponding to amino acid residues 421-431 of the human low density lipoprotein receptor-related protein (LRP) is completely conserved between LRP-5 and LRP-6, as well as between human and mouse.Specifications
LRP-5/6 | |
1A12 | |
In 150mM NaCl, 10mM Tris-glycine, 25% glycerol, 0.5% BSA, pH 7.0. | |
Murine | |
IgG2b | |
RUO | |
Primary | |
Recognizes the ∽175kDa full-length LRP-6 protein and a ∽145kDa proteolytic fragment. |
Immuno Blotting, Immunoblot | |
Unconjugated | |
Purified | |
a synthetic peptide (DTGTDRIEVTR) corresponding to amino acids 421-431 of the human low density lipoprotein receptor-related protein 6 (LRP-6) | |
50μg | |
≤ -70°C | |
Monoclonal | |
Human, Murine |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only