Learn More
LRP8, Mouse, Clone: 1A1, Abnova™
Mouse monoclonal antibody raised against a partial recombinant LRP8.
Supplier: Abnova Corporation H00007804M02
Description
This gene encodes an apolipoprotein E receptor, a member of the low density lipoprotein receptor (LDLR) family. Apolipoprotein E is a small lipophilic plasma protein and a component of lipoproteins such as chylomicron remnants, very low density lipoprotein (VLDL), and high density lipoprotein (HDL). The apolipoprotein E receptor is involved in cellular recognition and internalization of these lipoproteins. Alternative splicing generates multiple transcript variants encoding distinct isoforms for this gene. [provided by RefSeq
Sequence: KKTCADSDFTCDNGHCIHERWKCDGEEECPDGSDESEATCTKQVCPAEKLSCGPTSHKCVPASWRCDGEKDCEGGADEAGCATLCAPHSpecifications
LRP8, | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_004631 | |
LRP8 | |
LRP8 (NP_004622, 83 a.a. ∼ 170 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA, Western Blot | |
1A1 | |
Mouse monoclonal antibody raised against a partial recombinant LRP8. | |
LRP8 | |
APOER2/HSZ75190/MCI1 | |
Mouse | |
Affinity chromatography | |
RUO | |
7804 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.