Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-LRRC16A, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP191222

Catalog No. NBP191222


Add to cart

Description

Description

LRRC16A Polyclonal antibody specifically detects LRRC16A in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

LRRC16A
Unconjugated
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Arp2/3, and Myosin-I Linker homolog 1, CARMIL, CARMIL1, CARMIL1a, dJ501N12.1, dJ501N12.5, FLJ20048, FLJ43708, leucine rich repeat containing 16, leucine rich repeat containing 16A, leucine-rich repeat-containing protein 16A, LRRC16
Rabbit
IgG
0.1 ml
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:1000 - 1:2500
Affinity Purified
LRRC16A
This antibody was developed against Recombinant Protein corresponding to amino acids:HKLADHFSRRGKTLPQQESLEIELAEEKPVKRSIITVEELTEIERLEDLDTCMMTPKSKRKSIHSRMLRPVSRAFEMEFDL
Immunogen affinity purified
RUO
Primary
55604
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only