Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRRC34 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524101

Catalog No. PIPA524101


Add to cart

Specifications

Specifications

LRRC34
Polyclonal
PBS with 0.09% sodium azide
Q8IZ02, Q9DAM1
leucine-rich repeat-containing protein 34
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
151827, 71827
Western Blot
Unconjugated
LRRC34
Liquid
LRRC34
KLH conjugated synthetic peptide between 154-182 amino acids from the central region of human LRRC34
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit