Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRRC4 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526538

Catalog No. PIPA526538


Add to cart

Description

Description

This antibody is predicted to react with mouse and rat based on sequence homology.

The exact function of LRRC4 is unknown.
Specifications

Specifications

LRRC4
Polyclonal
PBS with 0.09% sodium azide
Q9HBW1
NGL-2, brain tumor associated protein LRRC4, brain tumor-associated protein BAG, leucine-rich repeat-containing 4, leucine-rich repeat-containing protein 4, nasopharyngeal carcinoma-associated gene 14 protein, netrin-G2 ligand
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
64101
Immunohistochemistry, Western Blot
Unconjugated
LRRC4
Liquid
LRRC4
KLH conjugated synthetic peptide between 531-559 amino acids from the C-terminal region of human LRRC4
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit