Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRRC4B Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523529

Catalog No. PIPA523529

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

LRRC4B
Polyclonal
PBS with 0.09% sodium azide
Q9NT99, P0C192
HSM, LRIG4, NGL-3, leucine-rich repeat-containing protein 4B, leucine-rich repeats and immunoglobulin-like domains 4, netrin-G3 ligand
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
272381, 94030
Western Blot
Unconjugated
LRRC4B
Liquid
LRRC4B
KLH conjugated synthetic peptide between 81-110 amino acids from the N-terminal region of human LRRC4B
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit