Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRRC57 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA524038
Description
This antibody is predicted to react with rat based on sequence homology.
LRRC57 contains 7 LRR (leucine rich repeats). However, the exact function remains unknown.Specifications
LRRC57 | |
Polyclonal | |
LRRC57 | |
leucine-rich repeat-containing protein 57 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
255252 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry, Western Blot | |
Unconjugated | |
Q8N9N7 | |
LRRC57 | |
KLH conjugated synthetic peptide between 198-227 amino acids from the C-terminal region of human LRRC57 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction