Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRRK2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA146443

Catalog No. PA146443


Add to cart

Description

Description

The target sequence has 88% sequence homology with mouse and rat. Suggested positive control: human brain lysate.

The novel LRRK2 gene (also known as PARK8), is responsible for up to 6% of all autosomal domit Parkinson ft.s disease (PD). LRRK2 missense mutations segregate with PARK8-linked PD and are implicated in other neurodegenerative diseases involving alpha-synuclein and tau pathology.
Specifications

Specifications

LRRK2
Polyclonal
Unconjugated
LRRK2
Liquid
LRRK2
Synthetic peptide corresponding to residues N(2506)LEKHIEVRKELAEKMR(2522) of human LRRK2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q5S007
AURA17, DARDARIN, PARK8, RIPK7, ROCO2
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
120892
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit