Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LSM4, Polyclonal, Invitrogen™

Catalog No. PIPA128314
Change view
Click to view available options
Quantity:
50 μg
Catalog No. Quantity
PIPA128314 50 μg
1 options

Catalog No. PIPA128314

Supplier: Thermo Scientific PA128314

Chicken Polyclonal Antibody

Sm-like proteins were identified in a variety of organisms based on sequence homology with the Sm protein family (see SNRPD2; MIM 601061). Sm-like proteins contain the Sm sequence motif, which consists of 2 regions separated by a linker of variable length that folds as a loop. The Sm-like proteins are thought to form a stable heteromer present in tri-snRNP particles, which are important for pre-mRNA splicing.

PA1-28314 detects LSM4 from mouse, human, rat samples.PA1-28314 has been successfully used in Western blot applications.The PA1-28314 immunogen is: Recombinant fragment, corresponding to amino acids 1-139 of Human LSM4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LSM4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Gene LSM4
Gene Accession No. Q9Y4Z0, Q9QXA5
Gene Alias YER112W
Gene Symbols LSM4
Host Species Chicken
Immunogen Recombinant fragment, corresponding to amino acids 1-139 of Human LSM4.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25804
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.