Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LSP-1 Mouse anti-Human, Unlabeled, Clone: 16, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610734

Catalog No. BDB610734


Add to cart

Description

Description

LSP-1 (Lymphocyte Specific Protein-1) is expressed in lymphocytes (B cells, cytotoxic T cells, and helper T cells), neutrophils, and macrophages and is involved in lymphocyte signal transduction. LSP-1 is a cytoplasmic membrane-bound protein that, following activation, translocates to the cytosol. This activation and translocation is mediated by phosphorylation of serine and threonine residues in the C-terminus of LSP-1 in response to concanavalin A or phorbal myristate acetate stimulation of murine T cells. LSP-1 contains two N-terminal calcium-binding sites and has also been shown to bind to actin. These results suggest that LSP-1 may be involved in the binding of actin to the plasma membrane following stimulation of PKC in murine T cells. The mRNA is constitutively expressed; however, the protein is only expressed following activation of the cell. Furthermore, LSP-1 is a substrate for MAPKAP kinase 2 (MAP kinase-activated protein kinase 2) in neutrophils.
Specifications

Specifications

LSP-1
16
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Western Blot
250μg/mL
Lymphocyte Specific Protein-1
Affinity Purified
Human LSP-1 aa. 75-215
50μg
Cell Biology
Primary
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit