Learn More
LSP1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA525357
Catalog No. PIPA525357
Add to cart
Description
Description
LSP1 is an intracellular F-actin bindingprotein. The protein is expressed in lymphocytes, neutrophils,macrophages, and endothelium and may regulate neutrophil motility,adhesion to fibrinogen matrix proteins, and transendothelialmigration. Alternative splicing results in multiple transcriptvariants encoding different isoforms.
Specifications
Specifications
LSP1
Polyclonal
Unconjugated
LSP1
Liquid
LSP1
KLH conjugated synthetic peptide between 108-134 amino acids from the central region of human LSP1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Flow Cytometry, Immunohistochemistry, Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
P33241
WP34, pp52, 47 kDa actin binding protein, 47 kDa actin-binding protein, 52 kDa phosphoprotein, F-actin binding and cytoskeleton associated protein, leufactin (leukocyte F-actin binding protein), leukocyte-specific protein 1, lymphocyte-specific antigen WP34
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
4046