Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LTBR, Polyclonal, Invitrogen™

Catalog No. PIPA127537
Change view
Click to view available options
Quantity:
50 μg
Catalog No. Quantity
PIPA127537 50 μg
1 options

Catalog No. PIPA127537

Supplier: Thermo Scientific PA127537

Chicken Polyclonal Antibody

Receptor for the heterotrimeric lymphotoxin containing LTA and LTB. and for TNFS14/LIGHT. Promotes apoptosis via TRAF3 and TRAF5. May play a role in the development of lymphoid organs. The protein encoded by this gene is a member of the tumor necrosis fac

PA1-27537 detects LTBR from human samples.PA1-27537 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The immunogen is a synthetic peptide corresponding to human Tumor necrosis factor receptor superfamily member 3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LTBR
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Gene LTBR
Gene Accession No. P36941
Gene Alias CD18, D12S370, LT-BETA-R, TNF-R-III, TNFCR, TNFR-RP, TNFR2-RP, TNFRSF3
Gene Symbols LTBR
Host Species Chicken
Immunogen Synthetic peptide corresponding to human Tumor necrosis factor receptor superfamily member 3.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4055
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.