Learn More
TRPM7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA528101
Description
PA5-28101 targets LTRPC7 in WB applications and shows reactivity with Human samples. The PA5-28101 immunogen is synthetic peptide corresponding to a region within amino acids 1802 and 1865 of Human LTRPC7. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
TRPCs, mammalian homologs of the Drosophila transient receptor potential (trp) protein, are ion channels that are thought to mediate capacitative calcium entry into the cell. TRP-PLIK is a protein that is both an ion channel and a kinase. As a channel, it conducts calcium and monovalent cations to depolarize cells and increase intracellular calcium. As a kinase, it is capable of phosphorylating itself and other substrates. The kinase activity is necessary for channel function, as shown by its dependence on intracellular ATP and by the kinase mutants.Specifications
TRPM7 | |
Polyclonal | |
Unconjugated | |
Q96QT4 | |
TRPM7 | |
Synthetic peptide corresponding to a region within amino acids 1802 and 1865 of Human LTRPC7 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
TRPM7 | |
CHAK, CHAK1, LTRPC7, TRP-PLIK, LTRPC ion channel family member 7, LTrpC-7, channel-kinase 1, long transient receptor potential channel 7, transient receptor potential cation channel subfamily M member 7, transient receptor potential-phospholipase C-interacting kinase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54822 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.