Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LUC7L2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526556

Catalog No. PIPA526556


Add to cart

Description

Description

This antibody is predicted to react with mouse based on sequence homology.

LUC7L2 may bind to RNA via its Arg/Ser-rich domain.
Specifications

Specifications

LUC7L2
Polyclonal
PBS with 0.09% sodium azide
Q9Y383
CGI-74, LUC7B2, putative RNA-binding protein Luc7-like 2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
51631
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
LUC7L2
Liquid
LUC7L2
KLH conjugated synthetic peptide between 341-369 amino acids from the C-terminal region of human LUC7L2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit