Learn More
l luciferase Rabbit anti-Bacteria, Luciferase, Biotin, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA126227
Catalog No. PA126227
Add to cart
Description
Description
PA1-26227 detects bacterial Luciferase from Photobacterium fischerii samples. PA1-26227 has been successfully used in Western blot, ELISA, immunocytochemistry, and immunomicroscopy procedures. The PA1-26227 immunogen is Luciferase from Photobacterium fischerii .
Luciferase is a generic name for an enzyme that catalyzes a light-emitting reaction. Luciferases can be found in bacteria, algae, fungi, jellyfish, insects, shrimp, and squid, and the resulting light that these organisms produce is termed bioluminescence. In bacteria, the genes responsible for the light-emitting reaction (the lux genes) have been isolated and used extensively in the construction of bioreporters that emit a blue-green light with a maximum intensity at 490 nm. Three variants of lux are available. The lux genetic system consists of five genes, luxA, luxB, luxC, luxD, and luxE. Depending on the combination of these genes used, several different types of bioluminescent bioreporters can be constructed.
Specifications
Specifications
Bacterial Luciferase
Polyclonal
Biotin
Liquid
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
Bacteria, Luciferase
ELISA, Immunocytochemistry, Immunofluorescence, Immunomicroscopy, Western Blot
10 mg/mL
0.02M potassium phosphate with 0.15M NaCl, 1% BSA, 50% glycerol and 0.01% sodium azide; pH 7.2
Lux
Luciferase from Photobacterium fischerii .
IgG fraction
RUO
Antibody
Polyclonal