Learn More
Luciferase Mouse anti-All, Clone: clone21, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05603
Catalog No. 05-603-MI
Add to cart
Description
Description
Detect Luciferase using this Anti-Luciferase Antibody, clone21 validated for use in western blot, immunohistochemistry.
Specifications
Specifications
Luciferase
21
0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Murine
IgG1
200μg
Primary
All
Immunohistochemistry, Western Blot
Unconjugated
Purified
Full-length recombinant luciferase from Photinus pyralis.
Protein G chromatography
−20°C for 24 months.
Monoclonal