Learn More
LHR Rabbit anti-Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA185662
Catalog No. PA185662
Add to cart
Description
Description
Reconstitute with distilled water. For prolonged storage, the addition of a preservative is recommended. Prior to reconstitution, store at 4°C. After reconstitution, store undiluted at -20°C, avoiding freeze/thaw cycles.
Specifications
Specifications
LHR
Polyclonal
0.05M Ammonium bicarbonate with no preservative
P16235
LH receptor, Lhr
Rabbit
IgG
50 μL
4° C
Primary
25477
Western Blot
Unconjugated
Lhcgr
Lyophilized
LHCGR
Synthetic peptide corresponding to a C-terminal portion of the native molecule, conjugated to thyroglobulin.
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Rat