Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LUZP1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524647

Catalog No. PIPA524647

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

LUZP1 encodes a protein that contains a leucine zipper motif. The exact function of the encoded protein is not known. In mice this gene affects neural tube closure.
Specifications

Specifications

LUZP1
Polyclonal
PBS with 0.09% sodium azide
Q86V48
LUZP
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
7798
Flow Cytometry, Western Blot
Unconjugated
LUZP1
Liquid
LUZP1
KLH conjugated synthetic peptide between 1042-1070 amino acids from the C-terminal region of human LUZP1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit