Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Latexin, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen PA129870

Catalog No. PIPA129870

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene encodes the only known protein inhibitor of zinc-dependent metallocarboxypeptidases.

PA1-29870 detects LXN from rat, human, mouse samples.PA1-29870 has been successfully used in immunocytochemistry, immunohistochemistry applications.The PA1-29870 immunogen is: Recombinant fragment, corresponding to amino acids 1-222 of Human LXN.
Specifications

Specifications

LXN
Polyclonal
Unconjugated
LXN
Purified
LXN
Recombinant fragment, corresponding to amino acids 1-222 of Human LXN.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Murine, Rat
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry
1mg/mL
PBS with no preservative
Q9BS40, P70202, Q64361
ECI, TCI
Chicken
IgY
50μg
Store at -20°C
Primary
56925
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit