Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LXR alpha, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA1332

Catalog No. PA1332

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Liver X receptors (LXRs), also known as RLD1, are oxysterol activated nuclear receptors which are involved in the regulation of genesis and metabolism of cholesterols and bile acids. LXRs form heterodimeric complexes with the retinoic acid receptors (RARs) once activated by their respective oxysterol ligands. Two known isoforms, alpha and beta, are differentially expressed; the alpha isoform predominantly expressed in liver, whereas beta isoform expression has been shown to be ubiquitous.The corresponding gene for the LXR alpha is NR1H3.

PA1-332 detects LXR alpha in mouse samples. PA1-332 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~50kDa protein representing LXR alpha in mouse liver samples.The PA1-332 immunogen is a synthetic peptide corresponding to residues T(35) C I L R E E A R M P Q S T G(49) of mouse LXR alpha. The PA1-332 immunizing peptide (Cat. # PEP-272) is available for use in neutralization and control experiments.
Specifications

Specifications

LXR alpha
Polyclonal
Unconjugated
Nr1h3
Liquid
NR1H3
Synthetic peptide corresponding to residues T(35) C I L R E E A R M P Q S T G(49) of mouse LXR alpha.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Murine
Western Blot
1mg/mL
PBS with 1mg/mL BSA and 0.05% sodium azide
Q9Z0Y9
Liver X Receptor Alpha, Oxysterols receptor LXR-alpha, Liver X receptor alpha, Nuclear orphan receptor LXR-alpha
Rabbit
IgG
100μg
-20° C, Avoid Freeze/Thaw Cycles, Avoid Freeze/Thaw Cycles
Primary
22259
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit