Learn More
Ly-6A/E Rat anti-Mouse, BUV395, Clone: D7, BD
Rat Monoclonal Antibody
Supplier: BD Biosciences 563990
Description
The D7 antibody reacts with Ly-6A.2 and Ly-6E.1, which are allelic members of the Ly-6 multigene family. Sca-1 (Ly6A/E), a phosphatidylinositol-anchored protein of about 18 kDa, is expressed on the multipotent hematopoietic stem cells (HSC) in the bone marrow of mice with both Ly-6 haplotypes. In mice expressing the Ly-6.2 haplotype (e.g., AKR, C57BL, C57BR, C57L, C58, DBA/2, PL, SJL, SWR, 129), Ly-6A/E is also expressed on distinct subpopulations of bone marrow and peripheral B lymphocytes and thymic and peripheral T lymphocytes. Strains with the Ly-6.1 haplotype (e.g., A, BALB/c, CBA, C3H/He, DBA/1, NZB) have few Ly-6A/E+ resting peripheral lymphocytes, whereas activation of lymphocytes from mice of both Ly-6 haplotypes leads to strong expression of the Sca-1 antigen. Studies with the D7 antibody have demonstrated that Ly-6A/E may be involved in the regulation of B and T lymphocyte responses, and it appears to be required for T-cell receptor-mediated T-cell activation. Purified E13-161.7 mAb (anti-Ly-6A/E, Cat. No. 553333) can block binding of FITC-conjugated D7 antibody (Cat. No. 557405) to mouse splenocytes, but purified mAb D7 is unable to block binding of FITC-conjugated E13-161.7 antibody (Cat. No. 553335). Anti-Ly-6A/E (Sca-1) mAb may be used in combination with the Mouse Lineage Panel (Cat. No. 559971) to identify HSC.
The antibody was conjugated to BD Horizon™ BUV395 which has been exclusively developed by BD Biosciences as an optimal dye for use on a 355nm laser equipped instrument. With an Ex Max at 348 nmand an Em Max at 395nm, this dye has virtually no spillover into any other detector. BD Horizon™ BUV395 can be excited with a 355nm laser and detected with a 379/28 filter.
Specifications
Ly-6A/E | |
Monoclonal | |
0.2mg/mL | |
Ly-6A/E; Lymphocyte antigen 6A-2/6E-1; Ly-6A.2/Ly-6E.1; Sca-1; TAP | |
Rat | |
Affinity Purified | |
RUO | |
Mouse | |
IgG2a κ |
Flow Cytometry | |
D7 | |
Brilliant Ultraviolet 395 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
IL-2-dependent mouse T-cell line CTL-L | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.