Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Ly-6G FITC, Clone: RB6-8C5,

Rat Monoclonal Antibody

$105.70 - $153.50

Specifications

Antigen Ly-6G
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Format Purified
Formulation Purified Rat monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2.
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABF1481MI
View Documents
MilliporeSigma™
MABF1481
25μg
Each for $153.50
Add to cart
 
MABF1482MI
View Documents
MilliporeSigma™
MABF1482
100μg
Each for $105.70
Add to cart
 
Description

Description

Specifically detects Ly-6G Clone: RB6-8C5 in Mouse samples, and it is validated for Flow Cytometry

The RB6-8C5 antibody binds to mouse Ly-6G, commonly known as Gr-1, a member of the Ly-6 superfamily of GPI-anchored cell surface proteins with roles in cell signaling and cell adhesion. Gr-1 is differentially expressed during development and maturation of cells in the myeloid lineage and is expression at varying stages and levels on monocytes, macrophages, granulocytes, and peripheral neutrophils. In the mouse, the RB6-8C5 antibody is typically used in combination with the macrophage labeling antibody M1/70 (Anti-CD11b) for phenotypic analysis of monocytes, macrophages and granulocytes.Note: The RB6-8C5 antibody has been reported to cross-react with Ly-6C on cells expressing this antigen (Fleming et al. 1993. J. Immunol. 151:2399-2408 and Sasmono et al. 2007. J. Leukoc. Biol. 82: 111-123) and has been cited in the literature for identification of Ly-6G/Ly-6C. Other reports suggest that this antibody is specific for Ly-6G, without cross-reactivity for Ly-6C (Nagendra S. and Schlueter AJ. 2003. Cytometry A, 58(2): 195-200.
Specifications

Specifications

Ly-6G
Monoclonal
Purified Rat monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2.
Flow Cytometry
Proprietary immunogen
Ly6g
RUO
Purified by size-exclusion chromatography
Primary
Purified
RB6-8C5
Rat
P35461
2°C to 8°C undiluted for one year from date of shipment protected from light; Do not freeze
Inflammation, Immunology
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit