Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-Ly-6G, PE, Clone: 1A8,
Rat Monoclonal Antibody
$299.00
Specifications
Antigen | Ly-6G |
---|---|
Clone | 1A8 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | PE |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABF1423MI
|
MilliporeSigma™
MABF1423 |
100 μg |
Each of 1 for $299.00
|
|
|||||
Description
Specifically detects Ly-6G Clone: 1A8 in Mouse samples, and it is validated for Flow Cytometry
The 1A8 antibody binds to mouse Ly-6G, commonly known as Gr-1, a member of the Ly-6 superfamily of GPI-anchored cell surface proteins with roles in cell signaling and cell adhesion. Gr-1 is differentially expressed during development and maturation of cells in the myeloid lineage and is expression at varying stages and levels on monocytes, macrophages, granulocytes, and peripheral neutrophils. In the mouse, the 1A8 antibody is typically used in combination with the macrophage labeling antibody M1/70 (Anti-CD11b) for phenotypic analysis of monocytes, macrophages and granulocytes.Specifications
Ly-6G | |
Flow Cytometry | |
PE | |
Rat | |
Inflammation, Immunology | |
Purified Rat monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2. | |
Ly6g | |
IgG2a κ | |
Purified by size-exclusion chromatography |
1A8 | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
P35461 | |
Proprietary immunogen | |
Primary | |
2°C to 8°C for one year from date of shipment protected from light; Do not freeze |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title