Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LYAR Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA514213

Catalog No. PIPA514213

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

LYAR
Polyclonal
PBS with 0.09% sodium azide
Q9NX58
Cell growth-regulating nucleolar protein; PNAS-5; LYAR
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
55646
Immunohistochemistry, Western Blot
Unconjugated
LYAR
Liquid
LYAR
KLH conjugated synthetic peptide between 38-66 amino acids from the N-terminal region of human LYAR
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit