Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lyn Mouse anti-Human, Mouse, Rat, Chicken, Clone: 10A6.2, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 04268
Description
Anti-Lyn Antibody, clone 10A6.2 is an antibody against Lyn for use in western blotting.Specifications
Lyn | |
Monoclonal | |
Unconjugated | |
NP_001104567 | |
Mouse | |
100 μL | |
Chicken, Human, Mouse, Rat | |
Ascites |
Western Blot | |
10A6.2 | |
Mouse monoclonal IgG1κ ascites containing 0.05% sodium azide. | |
JTK8, Lyn, FLJ26625 | |
KLH conjugated linear peptide corresponding to the C-terminus of Lyn. | |
Primary | |
−20°C for up to 12 months. | |
IgG1 κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only