Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LYRM1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA520920

Catalog No. PIPA520920


Add to cart

Description

Description

A suggested positive control is human liver tissue lysate. PA5-20920 can be used with blocking peptide PEP-1034.

LYRM1 was one of many genes identified through a suppression subtractive hybridization comparing their expression in omental adipose tissue of obese patients compared to non-obese individuals. Further study confirms that both mRNA and protein levels of LYRM1 are higher in obese individuals. LYRM1 is widely expressed, with highest levels occurring in adipose and liver tissues. It is thought that LYRM1 promotes preadipocyte proliferation and can inhibit apoptosis of preadipocytes. Ectopic LYRM1 expression did not significantly affect adipogenesis, suggesting that LYRM1 can influence adipose tissue homeostasis by modulating the size of the preadipocyte pool.
Specifications

Specifications

LYRM1
Polyclonal
Unconjugated
LYRM1
Liquid
LYRM1
A 17 amino acid peptide near the center of human LYRM1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
1 mg/mL
PBS with 0.02% sodium azide
O43325, Q9CQB7, B2RYU8
A211C6.1; LYR motif-containing protein 1
Rabbit
IgG
100 μg
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Primary
365361, 57149, 73919
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit