Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LYRM4 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA524175
Description
Required for nuclear and mitochondrial iron-sulfur protein biosynthesis.Specifications
LYRM4 | |
Polyclonal | |
LYRM4 | |
C6orf149, CGI-203, ISD11, LYR motif-containing protein 4, homolog of yeast Isd11, mitochondrial matrix Nfs1 interacting protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57128 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Q9HD34 | |
LYRM4 | |
KLH conjugated synthetic peptide between 43-69 amino acids from the central region of human LYRM4 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction