Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Lysine (K)-specific Demethylase 4A/KDM4A/JMJD2A, Alexa Fluor 700, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP149602AF700

Catalog No. NBP149602Z


Add to cart

Description

Description

Lysine (K)-specific Demethylase 4A/KDM4A/JMJD2A Polyclonal antibody specifically detects Lysine (K)-specific Demethylase 4A/KDM4A/JMJD2A in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

Lysine (K)-specific Demethylase 4A/KDM4A/JMJD2A
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
Affinity Purified
KDM4A
A genomic peptide made to an internal region of the human JMJD2A protein (within residues 320-460). [Swiss-Prot O75164]
Immunogen affinity purified
RUO
Store at 4C in the dark.
Polyclonal
Human, Mouse
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
O75164
EC 1.14.11, EC 1.14.11.-, JHDM3AJMJD2, JmjC domain-containing histone demethylation protein 3A, JMJD2Alysine-specific demethylase 4A, jumonji domain containing 2, jumonji domain containing 2A, Jumonji domain-containing protein 2A, KIAA0677jumonji C domain-containing histone demethylase 3A, lysine (K)-specific demethylase 4A
Rabbit
IgG
0.1 ml
Chromatin Modifiers, Epigenetics
Primary
9682
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only