Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Lysine (K)-specific Demethylase 4C/KDM4C/JMJD2C, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$119.60 - $397.50

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A genomic peptide made to an internal region of the human JMJD2C protein (within residues 450-600). [Swiss-Prot Q9H3R0]
Antigen Lysine (K)-specific Demethylase 4C/KDM4C/JMJD2C
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP149600SS
View Documents
Novus Biologicals
NBP149600SS
0.025 ml
Each for $119.60
Add to cart
 
NBP149600
View Documents
Novus Biologicals
NBP149600
0.1 ml
Each for $397.50
Add to cart
 
Description

Description

Lysine (K)-specific Demethylase 4C/KDM4C/JMJD2C Polyclonal antibody specifically detects Lysine (K)-specific Demethylase 4C/KDM4C/JMJD2C in Human, Mouse samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

Rabbit
A genomic peptide made to an internal region of the human JMJD2C protein (within residues 450-600). [Swiss-Prot Q9H3R0]
Polyclonal
Primary
Western Blot, ChIP assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
23081
0.5 mg/mL
IgG
Lysine (K)-specific Demethylase 4C/KDM4C/JMJD2C
Affinity Purified
RUO
Q9H3R0, Q8VCD7
PBS and 50% Glycerol with 0.05% Sodium Azide
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit