Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KARS Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA527620
Description
Recommended positive controls: H1299. Predicted reactivity: Mouse (94%), Rat (93%), Zebrafish (85%), Xenopus laevis (88%), Chicken (86%), Rhesus Monkey (96%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
KARS Polyclonal antibody specifically detects KARS in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
KARS | |
Polyclonal | |
Unconjugated | |
KARS1 | |
AA589550; AL024334; AL033315; AL033367; CMTRIB; D8Ertd698e; D8Wsu108e; DFNB89; EC 6.1.1.6; KARS; Kars1; KARS2; KIAA0070; KRS; lysine tRNA ligase; lysine--tRNA ligase; LysRS; lysyl-tRNA synthetase; mKIAA0070 | |
Rabbit | |
Protein A | |
RUO | |
3735, 85305 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
2 mg/mL | |
PBS with 40% glycerol and 0.01% thimerosal; pH 7 | |
Q15046, Q99MN1 | |
KARS1 | |
Recombinant fragment corresponding to a region within amino acids 280 and 597 of Human Lysyl-tRNA synthetase | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction