Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LZTR1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA520902
Description
A suggested positive control is human heart tissue lysate. PA5-20902 can be used with blocking peptide PEP-1016.
LZTR1 Polyclonal antibody specifically detects LZTR1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blot.Specifications
LZTR1 | |
Polyclonal | |
Unconjugated | |
LZTR1 | |
1200003E21Rik; AI591627; AW550890; BTBD29; fj87c05; leucine zipper like transcription regulator 1; leucine-zipper-like transcription regulator 1; leucine-zipper-like transcriptional regulator 1; leucine-zipper-like transcriptional regulator, 1; Lztr1; LZTR-1; MGC21205; NS10; si:dkey-98f17.2; SWNTS2; Tcfl2; wu:fj87c05; zgc:158250 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
360745, 66863, 8216 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q8N653, Q9CQ33 | |
LZTR1 | |
A 14 amino acid peptide near the amino terminus of human LZTR1. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction