Learn More
Bio X Cell InVivoPlus anti-mouse 4-1BB (CD137), Clone: LOB12.3, Size: 25 MG
SDP
Supplier:  Bio X Cell BP0169R025MG
Encompass_Preferred
InVivoPlus anti-mouse 4-1BB (CD137), Clone: LOB12.3, Size: 25 MG, Isotype: rat IgG2a, Applications: in vivo activation of 4-1BB
Catalog No. 50-139-3609