Learn More
Bio X Cell InVivoMab anti-mouse B220, Clone: RA3.3A1/6.1, Size: 25 MG
SDP
Supplier:  Bio X Cell BE0067R025MG
Encompass_Preferred
InVivoMab anti-mouse B220, Clone: RA3.3A1/6.1, Size: 25 MG, Isotype: rat IgM, Applications: in vivo B cell depletion, in vitro B cell negative selection
Catalog No. 50-139-2861