Learn More
Bio X Cell InVivoMAb anti-mouse CD122 (IL-2Rβ), Clone: TM-Beta 1, Size: 5 MG
Supplier:  Bio X Cell BE0298R005MG
Encompass_Preferred
InVivoMAb anti-mouse CD122 (IL-2Rβ), Clone: TM-Beta 1, Size: 5 MG, Isotype: rat IgG2b, Applications: in vivo NK cell depletion, in vitro IL-2R blockade, Functional assays, Flow cytometry
Catalog No. 50-139-3477