Learn More
CD253 (TRAIL) Rat anti-Mouse, Clone: N2B2, eBioscience
Rat Monoclonal Antibody
Manufacturer: Invitrogen 14595181
Description
Description: The N2B2 monoclonal antibody reacts with mouse TNF-related apoptosis-inducing ligand (TRAIL), a member of the TNF superfamily. TRAIL is not detected on the surface of freshly isolated T, B, or NK cells, but can be induced preferentially on CD3^- NK1.1^+ NK cells after stimulation with IL-2 or IL-15. N2B2 inhibits IL-2- or IL-15-activated NK cell cytotoxicity against mouse fibrosarcoma L929 target cells. Applications Reported: N2B2 can be used in flow cytometric analysis, and blocking of TRAIL in functional assays. (Please use Functional Grade purified N2B2, cat. 16-5951, in functional assays.). Applications Tested: The N2B2 antibody has been tested by flow cytometric analysis of mouse TRAIL transfected cells and can be used at less than or equal to 1 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 µm post-manufacturing filtered.
Apoptosis, or programmed cell death, occurs during normal cellular differentiation and development of multicellular organisms. Apoptosis is induced by certain cytokines including TNF and Fas ligand in the TNF family through their death domain containing receptors, TNFR1 and Fas. A novel member in the TNF family was recently identified and designated TRAIL (for TNF-related apoptosis-inducing ligand) and Apo-2L (for Apo-2 ligand) (6,7). TRAIL is a type II membrane protein and expressed in a variety of human tissues. Two novel death domain containing receptors DR4 and DR5 have been identified as the receptor for TRAIL3-6. Like TNF and Fas ligand, TRAIL induces apoptosis and NF-kB activation in many tissues and cells.Specifications
CD253 (TRAIL) | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P50592 | |
Apo2L (for Apo-2 Ligand), TNF-Related Apoptosis-Inducing Ligand, Tumor necrosis factor ligand, superfamily member 10, TRAIL protein, CD253 antigen, TN | |
Rat | |
Affinity chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Mouse |
Flow Cytometry, Functional Assay, Immunoprecipitation, Neutralization | |
N2B2 | |
Unconjugated | |
Tnfsf10 | |
Liquid | |
Tnfsf10 | |
IgG2a, kappa | |
50 μg | |
4° C | |
Primary | |
22035 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
For Research Use Only.