Learn More
Bio X Cell InVivoMab anti-mouse CD3ε, Clone: 145-2C11, Size: 100 MG
SDP
Supplier:  Bio X Cell BE00011R100MG
Encompass_Preferred
InVivoMab anti-mouse CD3ε, Clone: 145-2C11, Size: 100 MG, Isotype: Armenian hamster IgG, Applications: in vivo T cell depletion, in vitro T cell stimulation/activation, Immunofluorescence, Flow cytometry
Catalog No. 50-139-2705