Learn More
Bio X Cell InVivoPlus anti-mouse CD3ε, Clone: 145-2C11, Size: 25 MG
Supplier:  Bio X Cell BP00011R025MG
Encompass_Preferred
InVivoPlus anti-mouse CD3ε, Clone: 145-2C11, Size: 25 MG, Isotype: Armenian hamster IgG, Applications: in vivo T cell depletion, in vitro T cell stimulation/activation, Immunofluorescence, Flow cytometry
Catalog No. 50-139-3505