Learn More
Bio X Cell InVivoMab anti-mouse CD3ε F(ab')2 fragment, Clone: 145-2C11 f(ab')2 Fragments, Size: 5 MG
SDP
Supplier:  Bio X Cell BE00011FABR005MG
Encompass_Preferred
InVivoMab anti-mouse CD3ε F(ab')2 fragment, Clone: 145-2C11 f(ab')2 Fragments, Size: 5 MG, Isotype: hamster IgG2b, Applications: in vivo T cell depletion
Catalog No. 50-139-2700