Learn More
Bio X Cell InVivoPlus anti-mouse CD40L (CD154), Clone: MR-1, Size: 100 MG
Supplier:  Bio X Cell BP00171R100MG
Encompass_Preferred
InVivoPlus anti-mouse CD40L (CD154), Clone: MR-1, Size: 100 MG, Isotype: Armenian hamster IgG, Applications: in vivo blocking of CD40/CD40L signaling, in vitro blocking of CD40/CD40L signaling
Catalog No. 50-139-3522