Learn More
Bio X Cell InVivoMab anti-mouse CD47 (IAP), Clone: MIAP301, Size: 100 MG
Supplier:  Bio X Cell BE0270R100MG
Encompass_Preferred
InVivoMab anti-mouse CD47 (IAP), Clone: MIAP301, Size: 100 MG, Isotype: rat IgG2a, kappa, Applications: in vivo CD47 blockade, Immunofluorescence
Catalog No. 50-139-3376