Learn More
Bio X Cell InVivoPlus anti-mouse CD8α, Clone: 53-6.72, Size: 25 MG
SDP
Supplier:  Bio X Cell BP00041R025MG
Encompass_Preferred
InVivoPlus anti-mouse CD8α, Clone: 53-6.72, Size: 25 MG, Isotype: rat IgG2a, Applications: in vivo CD8+ T cell depletion, Immunofluorescence, Flow cytometry
Catalog No. 50-139-3512