Learn More
Bio X Cell InVivoPlus anti-mouse CD8α, Clone: 2.43, Size: 25 MG
SDP
Supplier:  Bio X Cell BP0061R025MG
Encompass_Preferred
InVivoPlus anti-mouse CD8α, Clone: 2.43, Size: 25 MG, Isotype: rat IgG2b, Applications: in vivo CD8+ T cell depletion
Catalog No. 50-139-3557