Learn More
Bio X Cell InVivoMab anti-mouse CD8α, Clone: YTS169.4, Size: 50 MG
SDP
Supplier:  Bio X Cell BE0117R050MG
Encompass_Preferred
InVivoMab anti-mouse CD8α, Clone: YTS169.4, Size: 50 MG, Isotype: rat IgG2b, Applications: in vivo CD8+ T cell depletion
Catalog No. 50-139-2966