Learn More
Bio X Cell InVivoMab anti-mouse CD8β (Lyt 3.2), Clone: 53-5.8, Size: 25 MG
Supplier:  Bio X Cell BE0223R025MG
Encompass_Preferred
InVivoMab anti-mouse CD8β (Lyt 3.2), Clone: 53-5.8, Size: 25 MG, Isotype: rat IgG1, Applications: in vivo CD8+ T cell depletion, in vitro CD8 blockade, Immunofluorescence
Catalog No. 50-139-3250