Learn More
Bio X Cell InVivoMab anti-mouse CSF1R (CD115), Clone: AFS98, Size: 25 MG
Supplier:  Bio X Cell BP0213R025MG
Encompass_Preferred
InVivoMab anti-mouse CSF1R (CD115), Clone: AFS98, Size: 25 MG, Isotype: rat IgG2a, Applications: in vivo macrophage depletion, in vitro CSF-R1 neutralization, in vivo monocyte depletion, Flow cytometry
Catalog No. 50-139-3621