Learn More
Bio X Cell InVivoMab anti-mouse CXCR3 (CD183), Clone: CXCR3-173, Size: 5 MG
SDP
Supplier:  Bio X Cell BE0249R005MG
Encompass_Preferred
InVivoMab anti-mouse CXCR3 (CD183), Clone: CXCR3-173, Size: 5 MG, Isotype: Armenian hamster IgG, Applications: in vivo CXCR3 neutralization, Flow cytometry
Catalog No. 50-139-3324