Learn More
Bio X Cell InVivoPlus anti-mouse/human/rat/monkey/hamster/canine/bovine TGF-β, Clone: 1D11.16.8, Size: 100 MG
SDP
Supplier:  Bio X Cell BP0057R100MG
Encompass_Preferred
InVivoPlus anti-mouse/human/rat/monkey/hamster/canine/bovine TGF-β, Clone: 1D11.16.8, Size: 100 MG, Isotype: mouse IgG1, Applications: in vivo TGFβ neutralization, in vitro TGFβ neutralization
Catalog No. 50-139-3549